CAS RN 98400-69-2
CAS RN 98400-69-2
IUPAC Name:
tert-butyl N-(pyridin-4-yl)carbamate
Synonyms:
tert-butyl N-(pyridin-4-yl)carbamate
Molecular Weight (g/mol):
194.23
Molecular Formula:
C10H14N2O2
InChi Key:
DRZYCRFOGWMEES-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)NC1=CC=NC=C1
1
–
1
of
1
results
1
4-(Boc-amino)pyridine, 95%
CAS: 98400-69-2 Molecular Formula: C10H14N2O2 Molecular Weight (g/mol): 194.234 MDL Number: MFCD02179232 InChI Key: DRZYCRFOGWMEES-UHFFFAOYSA-N Synonym: 4-boc-amino pyridine,4-tert-butoxycarbonylamino pyridine,tert-butyl pyridin-4-ylcarbamate,tert-butyl n-pyridin-4-yl carbamate,tert-butyl n-4-pyridyl carbamate,carbamic acid, 4-pyridinyl-, 1,1-dimethylethyl ester,pyridin-4-yl-carbamic acid tert-butyl ester,pubchem20060,acmc-209sa9,ksc495m3f PubChem CID: 9990210 IUPAC Name: tert-butyl N-pyridin-4-ylcarbamate SMILES: CC(C)(C)OC(=O)NC1=CC=NC=C1