accessibility menu, dialog, popup

UserName

CAS RN 98206-09-8

HClNNHOO

CAS RN 98206-09-8

IUPAC Name: hydrogen 1-(2,3-dihydro-1,4-benzodioxin-5-yl)piperazine chloride
Synonyms: eltoprazine
Molecular Weight (g/mol): 256.73
Molecular Formula: C12H17ClN2O2
InChi Key: JFSOSUNPIXJCIX-UHFFFAOYSA-N
SMILES: [H+].[Cl-].C1CN(CCN1)C1=CC=CC2=C1OCCO2
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results