accessibility menu, dialog, popup

UserName

H3COOClFFF

CAS RN 98187-17-8

IUPAC Name: 4-methoxy-2-(trifluoromethyl)benzoyl chloride
Synonyms: 4-methoxy-2-(trifluoromethyl)benzoyl chloride
Molecular Weight (g/mol): 238.59
Molecular Formula: C9H6ClF3O2
InChi Key: MYGJNVNUYJLTOZ-UHFFFAOYSA-N
SMILES: COC1=CC(=C(C=C1)C(Cl)=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results