accessibility menu, dialog, popup

UserName

OHFFBr

CAS RN 98130-56-4

IUPAC Name: 2-bromo-4,6-difluorophenol
Synonyms: 2-bromo-4,6-difluorophenol
Molecular Weight (g/mol): 208.99
Molecular Formula: C6H3BrF2O
InChi Key: YDRZYJATKMBSJX-UHFFFAOYSA-N
SMILES: OC1=C(F)C=C(F)C=C1Br
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results