accessibility menu, dialog, popup

UserName

HOBOH

CAS RN 98-80-6

IUPAC Name: phenylboronic acid
Synonyms: phenyl boronic acid β-phenyl-boronic acid kyselina fenylborita acide phenylborique phenylboronate phenyl-boric acid phenylboric acid boric acid, phenyl- usaf BO-2
Molecular Weight (g/mol): 121.93
Molecular Formula: C6H7BO2
InChi Key: HXITXNWTGFUOAU-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC=C1
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)