accessibility menu, dialog, popup

UserName

CH3H3CCH3OH

CAS RN 98-54-4

IUPAC Name: 4-tert-butylphenol
Synonyms: 4-tert-butylphenol P-T-butylyphenol para-tertiary-butylphenol p-terc.butylfenol PTBP ucar butylphenol 4-T caswell no. 130E epa pesticide chemical code 064113 ucar butylphenol 4-T flake butylphen
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: QHPQWRBYOIRBIT-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(O)C=C1
Molecular Weight (g/mol) 
  • (5)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)