accessibility menu, dialog, popup

UserName

CAS RN 98-23-7

CH3H3CH3CCH3H3CCH3

CAS RN 98-23-7

IUPAC Name: 5-tert-butyl-1,2,3-trimethylbenzene
Synonyms: 5-tert-butyl-1,2,3-trimethylbenzene
Molecular Weight (g/mol): 176.3
Molecular Formula: C13H20
InChi Key: ZQVJKYPEIPJEIP-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(C)=C1C)C(C)(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results