accessibility menu, dialog, popup

UserName

OFFHNSHN

CAS RN 97963-62-7

IUPAC Name: 5-(difluoromethoxy)-2,3-dihydro-1H-1,3-benzodiazole-2-thione
Synonyms: 5-(difluoromethoxy)-1,3-dihydro-1,3-benzodiazole-2-thione
Molecular Weight (g/mol): 216.21
Molecular Formula: C8H6F2N2OS
InChi Key: HJMVPNAZPFZXCP-UHFFFAOYSA-N
SMILES: FC(F)OC1=CC=C2NC(=S)NC2=C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results