CAS RN 97914-59-5
IUPAC Name:
2-(difluoromethoxy)benzoate
Synonyms:
2-(difluoromethoxy)benzoate
Molecular Weight (g/mol):
187.12
Molecular Formula:
C8H5F2O3
InChi Key:
AGDOJFCUKQMLHD-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1=CC=CC=C1OC(F)F
1
–
1
of
1
results
1
2-(Difluoromethoxy)benzoic acid, 97%, Thermo Scientific™
CAS: 97914-59-5 Molecular Formula: C8H5F2O3 Molecular Weight (g/mol): 187.12 MDL Number: MFCD00236223 InChI Key: AGDOJFCUKQMLHD-UHFFFAOYSA-M Synonym: 2-difluoromethoxy benzoic acid,benzoic acid, 2-difluoromethoxy,2-difluoromethoxy-benzoic acid,rarechem al bo 1907,timtec-bb sbb029907,art-chem-bb b006808,benzoic acid,2-difluoromethoxy,2-carboxy-alpha,alpha-difluoroanisole,2-bis fluoranyl methoxy benzoic acid PubChem CID: 2063348 IUPAC Name: 2-(difluoromethoxy)benzoic acid SMILES: [O-]C(=O)C1=CC=CC=C1OC(F)F