CAS RN 97858-62-3
CAS RN 97858-62-3
IUPAC Name:
(2-methoxyphenyl)({2-[(2-methoxyphenyl)(phenyl)phosphanyl]ethyl})phenylphosphane
Synonyms:
dipamp
Molecular Weight (g/mol):
458.48
Molecular Formula:
C28H28O2P2
InChi Key:
QKZWXPLBVCKXNQ-UHFFFAOYNA-N
SMILES:
COC1=CC=CC=C1P(CCP(C1=CC=CC=C1)C1=CC=CC=C1OC)C1=CC=CC=C1
1
–
1
of
1
results
1
(S,S)-DIPAMP, 97%
CAS: 97858-62-3 Molecular Formula: C28H28O2P2 Molecular Weight (g/mol): 458.48 MDL Number: MFCD05863547 InChI Key: QKZWXPLBVCKXNQ-UHFFFAOYNA-N Synonym: 1,2-bis s-2-methoxyphenyl phenyl phosphino ethane,s,s-dipamp,unii-hs6f5ew3u6,hs6f5ew3u6,ethylenebis s-o-anisyl phenyl phosphine,s,s-+-1,2-bis 2-methoxyphenyl phenyl phosphino ethane,s,s-dipamp; s,s-ethylenebis 2-methoxyphenyl phenylphosphine,s-2-methoxyphenyl 2-s-2-methoxyphenyl phenyl phosphanyl ethyl phenylphosphane PubChem CID: 11419748 IUPAC Name: (S)-(2-methoxyphenyl)-[2-[(2-methoxyphenyl)-phenylphosphanyl]ethyl]-phenylphosphane SMILES: COC1=CC=CC=C1P(CCP(C1=CC=CC=C1)C1=CC=CC=C1OC)C1=CC=CC=C1