accessibility menu, dialog, popup

UserName

CAS RN 977-96-8

ICH3NNH3C(E/Z)

CAS RN 977-96-8

IUPAC Name: 1-ethyl-2-[(1-ethyl-1,2-dihydroquinolin-2-ylidene)methyl]quinolin-1-ium iodide
Synonyms: pseudoisocyanine iodide diethylcyanine iodide decynium-22
Molecular Weight (g/mol): 454.36
Molecular Formula: C23H23IN2
InChi Key: GMYRVMSXMHEDTL-UHFFFAOYSA-M
SMILES: [I-].CCN1C(=CC2=CC=C3C=CC=CC3=[N+]2CC)C=CC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)