accessibility menu, dialog, popup

UserName

H3COOCH3CH2

CAS RN 97-88-1

IUPAC Name: butyl 2-methylprop-2-enoate
Synonyms: butyl methacrylate polyisobutyl methacrylate
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: SOGAXMICEFXMKE-UHFFFAOYSA-N
SMILES: CCCCOC(=O)C(C)=C
Molecular Weight (g/mol) 
  • (4)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)