CAS RN 96624-52-1
IUPAC Name:
4-butyl-4'-propyl-1,1'-bi(cyclohexane)
Synonyms:
4-butyl-4'-propyl-1,1'-bi(cyclohexane)
Molecular Weight (g/mol):
264.5
Molecular Formula:
C19H36
InChi Key:
QYGWJCMCLQAINT-UHFFFAOYSA-N
SMILES:
CCCCC1CCC(CC1)C1CCC(CCC)CC1
1
–
1
of
1
results
1
4-n-Butyl-4'-n-propylbicyclohexyl, 97%, Thermo Scientific™
CAS: 96624-52-1 Molecular Formula: C19H36 Molecular Weight (g/mol): 264.50 MDL Number: MFCD20260224 InChI Key: QYGWJCMCLQAINT-UHFFFAOYSA-N Synonym: trans,trans-4-butyl-4'-propyl-1,1'-bicyclohexyl,1-butyl-4-4-propylcyclohexyl cyclohexane,1s,1's,4r,4'r-4-butyl-4'-propyl-1,1'-bi cyclohexane,4-n-butyl-4'-n-propylbicyclohexyl,4-propyl-4'-butyl-1,1'-bicyclohexane,1-butyl-1-4-propylcyclohexyl cyclohexane,4-butyl-4'-propyl-1,1'-bi cyclohexane,1,1'-bicyclohexyl,4-butyl-4'-propyl-, trans,trans,trans,trans-4-butyl-4-propyl-1,1-bicyclohexyl,4alpha-butyl-4'beta-propyl-1beta,1'alpha-bicyclohexane PubChem CID: 11086663 SMILES: CCCCC1CCC(CC1)C1CCC(CCC)CC1