accessibility menu, dialog, popup

UserName

FClO

CAS RN 96516-31-3

IUPAC Name: 2-chloro-3-fluorobenzaldehyde
Synonyms: 2-chloro-3-fluorobenzaldehyde
Molecular Weight (g/mol): 158.56
Molecular Formula: C7H4ClFO
InChi Key: PIZVRLVKXWEMGO-UHFFFAOYSA-N
SMILES: FC1=CC=CC(C=O)=C1Cl
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results