accessibility menu, dialog, popup

UserName

CAS RN 96489-71-3

CH3H3CCH3NNClOSCH3H3CCH3

CAS RN 96489-71-3

IUPAC Name: 2-tert-butyl-5-{[(4-tert-butylphenyl)methyl]sulfanyl}-4-chloro-2,3-dihydropyridazin-3-one
Synonyms: pyridaben C19H25ClN2OS sanmite
Molecular Weight (g/mol): 364.93
Molecular Formula: C19H25ClN2OS
InChi Key: DWFZBUWUXWZWKD-UHFFFAOYSA-N
SMILES: CC(C)(C)N1N=CC(SCC2=CC=C(C=C2)C(C)(C)C)=C(Cl)C1=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
Concentration 
  • (1)
Element 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results