accessibility menu, dialog, popup

UserName

H3COONNNN

CAS RN 96107-94-7

IUPAC Name: 5-(ethoxycarbonyl)-2H-1,2,3,4-tetrazol-2-ide
Synonyms: 5-(ethoxycarbonyl)-2H-1,2,3,4-tetrazol-2-ide
Molecular Weight (g/mol): 141.11
Molecular Formula: C4H5N4O2
InChi Key: SIXBOTTXUOEPQW-UHFFFAOYSA-M
SMILES: CCOC(=O)C1=N[N-]N=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)