accessibility menu, dialog, popup

UserName

CAS RN 961-07-9

IUPAC Name: 2-amino-9-[4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-3H-purin-6-one
Synonyms: deoxyguanosine
Molecular Weight (g/mol): 267.25
Molecular Formula: C10H13N5O4
InChi Key: YKBGVTZYEHREMT-UHFFFAOYNA-N
SMILES: NC1=NC(=O)C2=C(N1)N(C=N2)C1CC(O)C(CO)O1
Molecular Weight (g/mol) 
  • (2)
  • (10)
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (4)
Physical Form 
  • (3)
  • (4)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (8)
  • (1)
  • (2)
  • (2)
  • (10)
  • (3)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (4)
  • (3)
  • (4)
  • (1)