accessibility menu, dialog, popup

UserName

CAS RN 960128-64-7

IUPAC Name: 8-(diphenylphosphanyl)-1,2,3,4-tetrahydronaphthalen-1-amine
Synonyms: 8-(diphenylphosphanyl)-1,2,3,4-tetrahydronaphthalen-1-amine
Molecular Weight (g/mol): 331.4
Molecular Formula: C22H22NP
InChi Key: KTWOFTBMHDQUEE-UHFFFAOYNA-N
SMILES: NC1CCCC2=C1C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results