accessibility menu, dialog, popup

UserName

CH3OOBr

CAS RN 96-32-2

IUPAC Name: methyl 2-bromoacetate
Synonyms: methyl 2-bromoacetate
Molecular Weight (g/mol): 152.98
Molecular Formula: C3H5BrO2
InChi Key: YDCHPLOFQATIDS-UHFFFAOYSA-N
SMILES: COC(=O)CBr
Boiling Point 
  • (3)
  • (4)
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (4)
  • (3)
  • (4)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)