accessibility menu, dialog, popup

UserName

CH3NHOCl

CAS RN 96-30-0

IUPAC Name: 2-chloro-N-methylacetamide
Synonyms: acetamide, 2-chloro-N-methyl-
Molecular Weight (g/mol): 107.54
Molecular Formula: C3H6ClNO
InChi Key: HOZLOOPIXHWKCI-UHFFFAOYSA-N
SMILES: CNC(=O)CCl
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results