accessibility menu, dialog, popup

UserName

CAS RN 96-21-9

HOBrBr

CAS RN 96-21-9

IUPAC Name: 1,3-dibromopropan-2-ol
Synonyms: 1,3-dibromo-2-propanol
Molecular Weight (g/mol): 217.89
Molecular Formula: C3H6Br2O
InChi Key: KIHQZLPHVZKELA-UHFFFAOYSA-N
SMILES: OC(CBr)CBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results