accessibility menu, dialog, popup

UserName

CAS RN 96-11-7

BrBrBr

CAS RN 96-11-7

IUPAC Name: 1,2,3-tribromopropane
Synonyms: 1,2,3-tribromopropane glycerol tribromohydrin sym-tribromopropane C3H5br3 s-tribromopropane glyceryl tribromohydrin
Molecular Weight (g/mol): 280.79
Molecular Formula: C3H5Br3
InChi Key: FHCLGDLYRUPKAM-UHFFFAOYSA-N
SMILES: BrCC(Br)CBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results