accessibility menu, dialog, popup

UserName

NFFFN

CAS RN 95727-86-9

IUPAC Name: 5-(trifluoromethyl)pyridine-2-carbonitrile
Synonyms: 5-(trifluoromethyl)pyridine-2-carbonitrile
Molecular Weight (g/mol): 172.11
Molecular Formula: C7H3F3N2
InChi Key: WDSCJULUXJSJOX-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CN=C(C=C1)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)