accessibility menu, dialog, popup

UserName

CAS RN 95266-40-3

H3COOOOHO(?)

CAS RN 95266-40-3

IUPAC Name: ethyl 4-[cyclopropyl(hydroxy)methylidene]-3,5-dioxocyclohexane-1-carboxylate
Synonyms: trinexapac-ethyl
Molecular Weight (g/mol): 252.27
Molecular Formula: C13H16O5
InChi Key: RVKCCVTVZORVGD-UHFFFAOYSA-N
SMILES: CCOC(=O)C1CC(=O)C(=C(O)C2CC2)C(=O)C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results