CAS RN 952518-97-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
4-{7H-pyrrolo[2,3-d]pyrimidin-4-yl}-1H-pyrazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-{7H-pyrrolo[2,3-d]pyrimidin-4-yl}-1H-pyrazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.