accessibility menu, dialog, popup

UserName

CH3(S)SH2NNH2O

CAS RN 95211-68-0

IUPAC Name: (6S)-2-amino-6-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide
Synonyms: (6S)-2-amino-6-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide
Molecular Weight (g/mol): 210.3
Molecular Formula: C10H14N2OS
InChi Key: LSRQCZCDBRHPHQ-YFKPBYRVSA-N
SMILES: C[C@H]1CCC2=C(C1)SC(N)=C2C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results