accessibility menu, dialog, popup

UserName

CAS RN 952-97-6

ONOS

CAS RN 952-97-6

IUPAC Name: 1-nitro-4-(phenylsulfanyl)benzene
Synonyms: 1-nitro-4-(phenylsulfanyl)benzene
Molecular Weight (g/mol): 231.27
Molecular Formula: C12H9NO2S
InChi Key: RJCBYBQJVXVVKB-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(SC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results