accessibility menu, dialog, popup

UserName

CAS RN 95010-17-6

BOBOBOH2CCH2H2CN

CAS RN 95010-17-6

IUPAC Name: pyridine; triethenyl-1,3,5,2,4,6-trioxatriborinane
Synonyms: pyridine; triethenyl-1,3,5,2,4,6-trioxatriborinane
Molecular Weight (g/mol): 240.67
Molecular Formula: C11H14B3NO3
InChi Key: YLHJACXHRQQNQR-UHFFFAOYSA-N
SMILES: C1=CC=NC=C1.C=CB1OB(OB(O1)C=C)C=C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results