accessibility menu, dialog, popup

UserName

CAS RN 95-69-2

CH3ClH2N

CAS RN 95-69-2

IUPAC Name: 4-chloro-2-methylaniline
Synonyms: 4-chloro-o-toluidine
Molecular Weight (g/mol): 141.6
Molecular Formula: C7H8ClN
InChi Key: CXNVOWPRHWWCQR-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=CC=C1N
Quantity 
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results