accessibility menu, dialog, popup

UserName

CAS RN 94944-70-4

H3CONHN(R)OH(S)OH(R)OHOH

CAS RN 94944-70-4

IUPAC Name: 1-{5-[(1R,2S,3R)-1,2,3,4-tetrahydroxybutyl]-1H-imidazol-2-yl}ethan-1-one
Synonyms: 1-{4-[(1R,2S,3R)-1,2,3,4-tetrahydroxybutyl]-3H-imidazol-2-yl}ethanone
Molecular Weight (g/mol): 230.22
Molecular Formula: C9H14N2O5
InChi Key: CQSIXFHVGKMLGQ-BWZBUEFSSA-N
SMILES: CC(=O)C1=NC=C(N1)[C@@H](O)[C@H](O)[C@H](O)CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results