accessibility menu, dialog, popup

UserName

CAS RN 947-19-3

HOO

CAS RN 947-19-3

IUPAC Name: 1-benzoylcyclohexan-1-ol
Synonyms: hydroxycyclohexyl phenyl ketone
Molecular Weight (g/mol): 204.27
Molecular Formula: C13H16O2
InChi Key: QNODIIQQMGDSEF-UHFFFAOYSA-N
SMILES: OC1(CCCCC1)C(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results