accessibility menu, dialog, popup

UserName

CAS RN 946409-38-7

NNNOOFFFFF

CAS RN 946409-38-7

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 2-(pyrrolidin-1-yl)pyrimidine-5-carboxylate
Synonyms: 2,3,4,5,6-pentafluorophenyl 2-(pyrrolidin-1-yl)pyrimidine-5-carboxylate
Molecular Weight (g/mol): 359.26
Molecular Formula: C15H10F5N3O2
InChi Key: YXDSLCBLWCPTJQ-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(OC(=O)C2=CN=C(N=C2)N2CCCC2)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results