accessibility menu, dialog, popup

UserName

CH3CH3HNSOO

CAS RN 94594-90-8

IUPAC Name: 10,10-dimethyl-4λ⁶-thia-3-azatricyclo[5.2.1.0²,⁶]decane-4,4-dione
Synonyms: 10,10-dimethyl-4λ⁶-thia-3-azatricyclo[5.2.1.0²,⁶]decane-4,4-dione
Molecular Weight (g/mol): 215.31
Molecular Formula: C10H17NO2S
InChi Key: IHFYOCYXHWUKQJ-UHFFFAOYNA-N
SMILES: CC1(C)C2CCC1C1NS(=O)(=O)CC21
Molecular Weight (g/mol) 
  • (3)
  • (4)
  • (1)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)