accessibility menu, dialog, popup

UserName

BNOOH3CH3CCH3FFFK

CAS RN 945493-51-6

IUPAC Name: potassium {1-[(tert-butoxy)carbonyl]-1H-indol-2-yl}trifluoroboranuide
Synonyms: potassium [1-(tert-butoxycarbonyl)indol-2-yl]trifluoroboranuide
Molecular Weight (g/mol): 323.16
Molecular Formula: C13H14BF3KNO2
InChi Key: IIPRPASPOXMUTF-UHFFFAOYSA-N
SMILES: [K+].CC(C)(C)OC(=O)N1C2=CC=CC=C2C=C1[B-](F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results