accessibility menu, dialog, popup

UserName

BOOH3CH3CCH3H3COONH

CAS RN 943994-02-3

IUPAC Name: 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-1,4-benzoxazin-3-one
Synonyms: 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,4-dihydro-1,4-benzoxazin-3-one
Molecular Weight (g/mol): 275.11
Molecular Formula: C14H18BNO4
InChi Key: ZXOSNHPLTJAXSA-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC2=C(OCC(=O)N2)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results