CAS RN 943764-99-6
CAS RN 943764-99-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Acebilustat, MedChemExpress
MedChemExpress Acebilustat (CTX-4430) is a leukotriene A4 hydrolase inhibitor, used for an oral antiinflammatory drug.