accessibility menu, dialog, popup

UserName

H3COCH3H3CCH3

CAS RN 943-27-1

IUPAC Name: 1-(4-tert-butylphenyl)ethan-1-one
Synonyms: P-tert-butyl acetophenone
Molecular Weight (g/mol): 176.26
Molecular Formula: C12H16O
InChi Key: UYFJYGWNYQCHOB-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C=C1)C(C)(C)C
Molecular Weight (g/mol) 
  • (3)
  • (5)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)