accessibility menu, dialog, popup

UserName

NNBrBr

CAS RN 942206-15-7

IUPAC Name: 5,6'-dibromo-3,3'-bipyridine
Synonyms: 5,6'-dibromo-3,3'-bipyridine
Molecular Weight (g/mol): 313.98
Molecular Formula: C10H6Br2N2
InChi Key: IMGBTSYZPFNIKJ-UHFFFAOYSA-N
SMILES: BrC1=CN=CC(=C1)C1=CN=C(Br)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results