accessibility menu, dialog, popup

UserName

CAS RN 942119-32-6

OOHNNN

CAS RN 942119-32-6

IUPAC Name: 2-(pyridin-3-yl)-3H,6H,11H-anthra[1,2-d]imidazole-6,11-dione
Synonyms: 2-(pyridin-3-yl)-3H-anthra[1,2-d]imidazole-6,11-dione
Molecular Weight (g/mol): 325.33
Molecular Formula: C20H11N3O2
InChi Key: IOBRUMYBBWCOCR-UHFFFAOYSA-N
SMILES: O=C1C2=CC=CC=C2C(=O)C2=C3N=C(NC3=CC=C12)C1=CN=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results