accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CClFFF

CAS RN 942069-65-0

IUPAC Name: 2-[3-chloro-5-(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-[3-chloro-5-(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 306.52
Molecular Formula: C13H15BClF3O2
InChi Key: DSRHHGXMHRASQY-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(=CC(Cl)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results