accessibility menu, dialog, popup

UserName

CAS RN 942-06-3

ClOOOCl

CAS RN 942-06-3

IUPAC Name: 5,6-dichloro-1,3-dihydro-2-benzofuran-1,3-dione
Synonyms: 5,6-dichloro-2-benzofuran-1,3-dione
Molecular Weight (g/mol): 217
Molecular Formula: C8H2Cl2O3
InChi Key: ULSOWUBMELTORB-UHFFFAOYSA-N
SMILES: ClC1=CC2=C(C=C1Cl)C(=O)OC2=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results