accessibility menu, dialog, popup

UserName

CAS RN 941717-05-1

ONNNOOFFFFF

CAS RN 941717-05-1

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 2-(morpholin-4-yl)pyrimidine-5-carboxylate
Synonyms: 2,3,4,5,6-pentafluorophenyl 2-(morpholin-4-yl)pyrimidine-5-carboxylate
Molecular Weight (g/mol): 375.26
Molecular Formula: C15H10F5N3O3
InChi Key: SXCSIAUTNMKWKC-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(OC(=O)C2=CN=C(N=C2)N2CCOCC2)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results