accessibility menu, dialog, popup

UserName

CAS RN 941717-00-6

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 4-(3,5-dimethyl-1H-pyrazol-1-yl)benzoate
Synonyms: 2,3,4,5,6-pentafluorophenyl 4-(3,5-dimethylpyrazol-1-yl)benzoate
Molecular Weight (g/mol): 382.29
Molecular Formula: C18H11F5N2O2
InChi Key: GQQALUKKLLEHOR-UHFFFAOYSA-N
SMILES: CC1=NN(C(C)=C1)C1=CC=C(C=C1)C(=O)OC1=C(F)C(F)=C(F)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results