accessibility menu, dialog, popup

UserName

CAS RN 941716-90-1

CH3HNO

CAS RN 941716-90-1

IUPAC Name: methyl({[4-(phenoxymethyl)phenyl]methyl})amine
Synonyms: methyl({[4-(phenoxymethyl)phenyl]methyl})amine
Molecular Weight (g/mol): 227.31
Molecular Formula: C15H17NO
InChi Key: PSMWUOSOWNRPOJ-UHFFFAOYSA-N
SMILES: CNCC1=CC=C(COC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results