CAS RN 941716-84-3
CAS RN 941716-84-3
IUPAC Name:
methyl({[1-(6-methylpyrazin-2-yl)piperidin-3-yl]methyl})amine
Synonyms:
methyl({[1-(6-methylpyrazin-2-yl)piperidin-3-yl]methyl})amine
Molecular Weight (g/mol):
220.32
Molecular Formula:
C12H20N4
InChi Key:
CBRQUNBWUQBINE-UHFFFAOYNA-N
SMILES:
CNCC1CCCN(C1)C1=NC(C)=CN=C1
1
–
1
of
1
results
1
N-Methyl-[1-(6-methylpyrazin-2-yl)piperid-3-yl]methylamine, 97%, Thermo Scientific™
CAS: 941716-84-3 Molecular Formula: C12H20N4 Molecular Weight (g/mol): 220.32 MDL Number: MFCD09879920 InChI Key: CBRQUNBWUQBINE-UHFFFAOYSA-N Synonym: n-methyl-1-6-methylpyrazin-2-yl piperid-3-yl methylamine,methyl 1-6-methylpyrazin-2-yl piperidin-3-yl methyl amine,methyl 1-6-methylpyrazin-2-yl 3-piperidyl methyl amine,n-methyl-1-1-6-methylpyrazin-2-yl piperidin-3-yl methanamine PubChem CID: 24229603 IUPAC Name: N-methyl-1-[1-(6-methylpyrazin-2-yl)piperidin-3-yl]methanamine SMILES: CC1=CN=CC(=N1)N2CCCC(C2)CNC