accessibility menu, dialog, popup

UserName

CAS RN 94-53-1

OHOOO

CAS RN 94-53-1

IUPAC Name: 2H-1,3-benzodioxole-5-carboxylic acid
Synonyms: 2H-1,3-benzodioxole-5-carboxylic acid
Molecular Weight (g/mol): 166.13
Molecular Formula: C8H6O4
InChi Key: VDVJGIYXDVPQLP-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C2OCOC2=C1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)