accessibility menu, dialog, popup

UserName

CAS RN 94-05-3

CH3ONOOH3C(Z)

CAS RN 94-05-3

IUPAC Name: ethyl (2Z)-2-cyano-3-ethoxyprop-2-enoate
Synonyms: ethyl (2Z)-2-cyano-3-ethoxyprop-2-enoate
Molecular Weight (g/mol): 169.18
Molecular Formula: C8H11NO3
InChi Key: KTMGNAIGXYODKQ-SREVYHEPSA-N
SMILES: CCO\C=C(\C#N)C(=O)OCC
Boiling Point 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)