accessibility menu, dialog, popup

UserName

CAS RN 937795-92-1

CH3NNNCl

CAS RN 937795-92-1

IUPAC Name: 2-[3-(chloromethyl)piperidin-1-yl]-6-methylpyrazine
Synonyms: 2-[3-(chloromethyl)piperidin-1-yl]-6-methylpyrazine
Molecular Weight (g/mol): 225.72
Molecular Formula: C11H16ClN3
InChi Key: DTVYKEHYUMEUFH-UHFFFAOYNA-N
SMILES: CC1=CN=CC(=N1)N1CCCC(CCl)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results