CAS RN 935841-80-8
CAS RN 935841-80-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
N-[(1R,2R)-2-Hydroxy-3-[(2-methylpropyl)[(4-nitrophenyl)sulfonyl]amino]-1-(phenylmethyl)propyl]carbamic Acid 1,1-Dimethylethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-[(1R,2R)-2-Hydroxy-3-[(2-methylpropyl)[(4-nitrophenyl)sulfonyl]amino]-1-(phenylmethyl)propyl]carbamic Acid 1,1-Dimethylethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.