CAS RN 935-69-3
CAS RN 935-69-3
IUPAC Name:
7-methyl-7H-purin-6-amine
Synonyms:
7-methyladenine
Molecular Weight (g/mol):
149.16
Molecular Formula:
C6H7N5
InChi Key:
HCGHYQLFMPXSDU-UHFFFAOYSA-N
SMILES:
CN1C=NC2=NC=NC(N)=C12
1
–
1
of
1
results
1
7-Methyl-7H-purin-6-amine, TRC
CAS: 935-69-3 Molecular Formula: C6 H7 N5 Molecular Weight (g/mol): 149.153 Synonym: 7-Methyl-adenine,6-Amino-7-methylpurine,7-Methyladenine,N7-Methyladenine,NSC 7857 IUPAC Name: 7-methylpurin-6-amine SMILES: Cn1cnc2ncnc(N)c12